Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269585.3 | NP_001256514.1 | 1 - 2 | 287 | 135 | |
NM_001380804.1 | NP_001367269.1 | 1 - 2 | 247 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |