Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269671.3 | NP_001256600.1 | 6 - 7 | 840 | 76 | |
NM_001269672.3 | NP_001256601.1 | 6 - 7 | 827 | 76 | ||
NM_001269673.4 | NP_001256602.1 | 6 - 7 | 813 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |