Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269711.3 | NP_001256640.1 | 17 - 18 | 2687 | 104 | |
NM_001269712.1 | NP_001256641.1 | 8 - 9 | 980 | 104 | ||
GenBank mRNA | AF318614.2 | - | 17 - 18 | 2663 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |