Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129446.4 | NP_001122918.1 | 4 - 5 | 1109 | 81 | |
NR_070019.1 | 5 - 6 | 904 | 81 | |||
NR_070020.1 | 4 - 5 | 781 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |