Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383468.1 | 4 - 5 | 671 | 75 |
Gene Symbol: mdt-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380831.2 | NP_001368121.1 | 2 - 3 | 332 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |