Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-33D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074675.4 | NP_507076.1 | 3 - 4 | 593 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |