Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074924.1 | NP_507325.1 | 1 - 2 | 514 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |