Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383483.1 | 5 - 6 | 2083 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |