Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075429.8 | NP_507830.1 | 2 - 3 | 521 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |