Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028720.5 | NP_001023891.2 | 7 - 8 | 1917 | 78 | |
NM_001028721.5 | NP_001023892.1 | 4 - 5 | 1316 | 78 | ||
NM_001028723.4 | NP_001023894.2 | 6 - 7 | 1792 | 78 | ||
NM_001313065.3 | NP_001299994.1 | 6 - 7 | 1798 | 78 | ||
NM_001373210.2 | NP_001359648.1 | 8 - 9 | 2114 | 78 | ||
NM_075607.7 | NP_508008.4 | 7 - 8 | 1905 | 78 | ||
GenBank mRNA | AF364604.1 | - | 7 - 8 | 1920 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |