Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374981.3 | NP_001362097.1 | 12 - 13 | 3550 | 143 | |
NM_076103.6 | NP_508504.2 | 13 - 14 | 3572 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |