Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313445.3 | NP_001300374.1 | 7 - 8 | 1657 | 128 | |
NM_001313446.4 | NP_001300375.1 | 9 - 10 | 1954 | 128 | ||
NM_001380960.1 | NP_001368582.1 | 9 - 10 | 1933 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |