Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jkk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383579.1 | 7 - 8 | 924 | 99 | ||
GenBank mRNA | AB024086.1 | - | 7 - 8 | 864 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |