Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029203.3 | NP_001024374.1 | 3 - 4 | 495 | 108 | |
NM_001380987.1 | NP_001367469.1 | 3 - 4 | 378 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |