Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129635.6 | NP_001123107.2 | 3 - 4 | 817 | 70 | |
NM_076644.7 | NP_509045.1 | 4 - 5 | 1003 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |