Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029736.6 | NP_001024907.2 | 2 - 3 | 559 | 91 | |
NM_001029737.5 | NP_001024908.2 | 2 - 3 | 357 | 91 | ||
NM_001038472.5 | NP_001033561.1 | 2 - 3 | 445 | 91 | ||
NM_001381012.1 | NP_001368078.1 | 2 - 3 | 325 | 91 | ||
GenBank mRNA | AY485309.1 | - | 2 - 3 | 353 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |