Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076769.4 | NP_509170.1 | 2 - 3 | 280 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |