Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076892.7 | NP_509293.1 | 4 - 5 | 422 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |