Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029659.5 | NP_001024830.1 | 9 - 10 | 1584 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |