Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373414.1 | NP_001360736.1 | 2 - 3 | 435 | 115 | |
NM_001373415.1 | NP_001360737.1 | 2 - 3 | 435 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |