Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077165.7 | NP_509566.1 | 7 - 8 | 737 | 60 | |
GenBank mRNA | AF082010.1 | - | 7 - 8 | 737 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |