Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129669.4 | NP_001123141.2 | 30 - 31 | 4496 | 122 | |
NM_001373489.2 | NP_001360637.1 | 30 - 31 | 4475 | 122 | ||
NM_001373491.2 | NP_001360639.1 | 30 - 31 | 4505 | 122 | ||
NM_001373492.2 | NP_001360635.1 | 30 - 31 | 4526 | 122 | ||
NM_001373493.2 | NP_001360636.1 | 31 - 32 | 4646 | 122 | ||
NM_001373494.2 | NP_001359720.1 | 31 - 32 | 4667 | 122 | ||
NM_001373495.2 | NP_001360638.1 | 31 - 32 | 4676 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |