Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077543.5 | NP_509944.1 | 2 - 3 | 223 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |