Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sad-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083291.4 | NP_001076760.1 | 4 - 5 | 467 | 77 | |
NM_001083292.4 | NP_001076761.1 | 4 - 5 | 473 | 77 | ||
GenBank mRNA | AF316542.1 | - | 4 - 5 | 455 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |