Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cah-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077863.8 | NP_510264.1 | 3 - 4 | 232 | 108 | |
NM_077864.5 | NP_510265.1 | 3 - 4 | 362 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |