Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029441.4 | NP_001024612.1 | 7 - 8 | 1052 | 75 | |
NM_077956.8 | NP_510357.3 | 7 - 8 | 1052 | 75 | ||
GenBank mRNA | AF072381.1 | - | 7 - 8 | 1052 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |