Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ram-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078021.4 | NP_510422.2 | 15 - 16 | 1883 | 73 | |
GenBank mRNA | AF218866.1 | - | 15 - 16 | 1905 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |