Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067962.7 | NP_500363.1 | 3 - 4 | 420 | 152 | |
GenBank mRNA | AF226867.1 | - | 3 - 4 | 386 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |