Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hrpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047480.6 | NP_001040945.1 | 3 - 4 | 952 | 63 | |
NM_001380140.2 | NP_001367846.1 | 3 - 4 | 958 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |