Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002996.1 | NP_001002996.1 | 1 | 174 | 101 | |
XM_005627797.3 | XP_005627854.1 | 1 | 308 | 101 | ||
XM_005627798.3 | XP_005627855.1 | 1 | 383 | 101 | ||
XM_005627799.3 | XP_005627856.1 | 1 | 308 | 101 | ||
XM_022426491.1 | XP_022282199.1 | 1 | 263 | 101 | ||
GenBank mRNA | AB083366.1 | - | 1 | 174 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |