Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002981.1 | NP_001002981.1 | 3 - 4 | 630 | 146 | |
XM_014111303.2 | XP_013966778.1 | 5 - 6 | 573 | 146 | ||
XM_022415465.1 | XP_022271173.1 | 4 - 5 | 414 | 146 | ||
XM_022415466.1 | XP_022271174.1 | 5 - 6 | 617 | 146 | ||
GenBank mRNA | AF086711.1 | - | 3 - 4 | 351 | 146 | |
AY333790.1 | - | 3 - 4 | 630 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Inventoried Probe spans exons |