Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166047.2 | NP_001159519.2 | 8 - 9 | 1107 | 120 | |
XM_005636672.3 | XP_005636729.1 | 9 - 10 | 1208 | 120 | ||
XM_005636673.3 | XP_005636730.1 | 8 - 9 | 1150 | 120 | ||
XM_022410841.1 | XP_022266549.1 | 8 - 9 | 1395 | 120 | ||
XM_022410842.1 | XP_022266550.1 | 7 - 8 | 1298 | 120 | ||
GenBank mRNA | AF403244.1 | - | 3 - 4 | 451 | 120 | |
GQ497899.1 | - | 8 - 9 | 1107 | 120 | ||
LN833579.1 | - | 8 - 9 | 1077 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |