Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003063.1 | NP_001003063.1 | 3 - 4 | 489 | 90 | |
GenBank mRNA | AF448499.1 | - | 3 - 4 | 489 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |