Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003166.1 | NP_001003166.1 | 1 - 2 | 100 | 82 | |
GenBank mRNA | AJ238952.1 | - | 1 - 2 | 100 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Inventoried Probe spans exons |