Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003045.1 | NP_001003045.1 | 2 - 3 | 561 | 58 | |
GenBank mRNA | AJ243320.1 | - | 2 - 3 | 561 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |