Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003359.1 | NP_001003359.1 | 2 - 3 | 346 | 61 | |
GenBank mRNA | AY392089.1 | - | 2 - 3 | 346 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |