Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113456.1 | NP_001106927.1 | 130 | 96 | ||
XM_014107713.2 | XP_013963188.1 | 346 | 96 | |||
GenBank mRNA | AH014135.2 | - | 130 | 96 | ||
EU334661.1 | - | 130 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |