Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031821.1 | NP_001026991.1 | 188 | 78 | ||
GenBank mRNA | AY916759.1 | - | 188 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Canis familiaris |