Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048097.1 | NP_001041562.1 | 1 - 2 | 801 | 129 | |
GenBank mRNA | DQ138060.1 | - | 1 - 2 | 801 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris Probe spans exons |