Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003298.1 | NP_001003298.1 | 7 - 8 | 1884 | 108 | |
XM_005621304.2 | XP_005621361.1 | 7 - 8 | 1805 | 108 | ||
XM_005621305.3 | XP_005621362.1 | 7 - 8 | 1747 | 108 | ||
GenBank mRNA | U32086.1 | - | 7 - 8 | 1884 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |