Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003348.1 | NP_001003348.1 | 405 | 140 | ||
XM_022422541.1 | XP_022278249.1 | 1596 | 140 | |||
GenBank mRNA | AB180206.1 | - | 405 | 140 | ||
AY741504.1 | - | 389 | 140 | |||
KT693114.1 | - | 383 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |