Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284450.1 | NP_001271379.1 | 7 - 8 | 1504 | 78 | |
XM_022421972.1 | XP_022277680.1 | 7 - 8 | 962 | 78 | ||
XM_022422000.1 | XP_022277708.1 | 7 - 8 | 976 | 78 | ||
XM_022422022.1 | XP_022277730.1 | 7 - 8 | 1063 | 78 | ||
GenBank mRNA | AB635686.1 | - | 7 - 8 | 1504 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Inventoried Probe spans exons |