Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431514.3 | XP_003431562.2 | 4 - 5 | 504 | 77 | |
XM_003431515.4 | XP_003431563.2 | 4 - 5 | 592 | 77 | ||
XM_005625978.3 | XP_005626035.1 | 4 - 5 | 591 | 77 | ||
XM_005625979.3 | XP_005626036.1 | 4 - 5 | 595 | 77 | ||
XM_005625980.3 | XP_005626037.1 | 4 - 5 | 517 | 77 | ||
XM_005625981.3 | XP_005626038.1 | 4 - 5 | 482 | 77 | ||
XM_005625982.3 | XP_005626039.1 | 4 - 5 | 450 | 77 | ||
XM_022424540.1 | XP_022280248.1 | 4 - 5 | 437 | 77 | ||
XM_857845.5 | XP_862938.3 | 4 - 5 | 587 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |