Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534128.6 | XP_534128.3 | 1 - 2 | 3154 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |