Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639856.2 | XP_003639904.1 | 1 - 2 | 195 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |