Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635701.2 | XP_005635758.1 | 8 - 9 | 1341 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |