Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253787.1 | NP_001240716.1 | 5 - 6 | 1061 | 83 | |
GenBank mRNA | AY906958.1 | - | 5 - 6 | 1061 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |