Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535637.6 | XP_535637.3 | 11 - 12 | 1238 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |