Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847624.5 | XP_852717.1 | 2 - 3 | 334 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |