Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630495.2 | XP_005630552.1 | 3 - 4 | 1337 | 73 | |
XM_022404758.1 | XP_022260466.1 | 4 - 5 | 1295 | 73 | ||
XM_022404759.1 | XP_022260467.1 | 4 - 5 | 1262 | 73 | ||
XM_022404760.1 | XP_022260468.1 | 4 - 5 | 1262 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |